Learn More
Medchemexpress LLC L-Aspartic acid | 56-84-8 | 133.10 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN066610MM/1ML
Encompass_Preferred
L-Aspartic acid is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues and in the study of inflammatory conditions. It is also an endogenous metabolite.
  • Used for preparing prodrugs to target colon and cecal tissues
  • Utilized in the study of inflammatory conditions
  • Acts as an endogenous metabolite
  • Purity of 99.84%
  • Suitable for colon-specific drug delivery
  • Increases L-[3H]Asp in the brain
  • Increases L-[3H]Glu levels
  • Exhibits a faster efflux rate across the Blood-Brain Barrier compared to L-Glu
Catalog No. 50-003-88400