Learn More
Bioworld L-Aspartic Acid, 500 g
Supplier:  Bioworld 401002881
Encompass_Preferred
Synonym: (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid Specifications:High Purity GradeSpecific rotation (4%, 6N HCL): +24.5º to +26.0ºMoisture (105ºC, 3hr): <0.2%Ammonia: <0.020%Chloride: <0.021%Sulfate: <0.028%Heavy metals (as Pb): <0.001%Iron: <0.002%Appearance: White Crystalline PowderBoiling Point : 264.1 °CDensity : 1.51 g/cm3HS Code : 29224995IUPAC_Name: 2-Aminobutanedioic AcidMelting Point: 300 °CPH Value: 2.28RTECS Number: CI9098500
Catalog No. 50-255-290