Learn More
Medchemexpress LLC L-Aspartic acid-d3 | 3842-25-9 | 136.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN0666S850MG
Encompass_Preferred
L-Aspartic acid-d3 is the deuterium-labeled form of L-Aspartic acid, an amino acid capable of penetrating the blood-brain barrier. It is commonly used in preparing prodrugs for colon and cecal tissues and in studying inflammatory conditions. Deuteration, the incorporation of stable heavy isotopes like deuterium, can affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitative analysis.
  • Functions as an internal standard for NMR, GC-MS, or LC-MS.
  • Suitable for research on inflammatory conditions.
  • Can be used in prodrug development targeting specific tissues.
Catalog No. 50-003-88379