Learn More
Medchemexpress LLC L-Aspartic acid dibenzyl ester 4-toluenesulfonate | 2886-33-1 | MFCD00065188 | ≥ 99% | 485.55 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW013719
Encompass_Preferred
L-Aspartic acid dibenzyl ester 4-toluenesulfonate is a pale brown powder that melts between 157-160 °C. It is utilized in the preparation of Betulinic Acid derivatives, which exhibit inhibition towards HIV, and also in the synthesis of piperidine functional amides, known for their potent and highly selective human β3 agonist activity.
  • Pale brown powder
  • Melting point 157-160 °C
  • Used in preparation of Betulinic Acid derivatives
  • Used in preparation of piperidine functional amides
Catalog No. 50-004-54010