Learn More
Medchemexpress LLC L-aspartic acid β-hydroxamate | 1955-68-6 | MFCD00050389 | 98.0% | 148.12 g/mol | C4H8N2O4 | 5 MG

Supplier: Medchemexpress LLC HY1344505MG
L-Aspartic acid β-hydroxamate is an aspartic acid derivative supplied as a white to off-white solid for laboratory research. The compound is well characterized (C4H8N2O4, 148.12 g/mol) and is provided at high purity for biochemical and pharmacological studies.
- High purity (98.0%) suitable for research applications.
- White to off-white solid for convenient handling.
- Molecular weight 148.12 g/mol and formula C4H8N2O4.
- Soluble in water (62.5 mg/mL with warming); limited solubility in DMSO.
- Store powder at -20°C for long-term stability; shorter stability in solution.
- Intended for research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.