Learn More
Alkali Scientific L Aspartic Acid Monopotassium Salt, 500 Grams
SDP
Supplier:  Alkali Scientific SPA006CP
Encompass_Preferred
L-Aspartic Acid Monopotassium Salt is an amino acid derivative commonly used in biochemical and metabolic research. It plays a key role in cellular metabolism as an intermediate in the citric acid cycle and amino acid biosynthesis pathways. This compound is frequently used in enzymatic assays and metabolic studies to investigate nitrogen transfer and energy production mechanisms. The monopotassium salt form enhances solubility and stability in aqueous solutions, making it suitable for laboratory applications. It is also utilized in cell culture media formulations and biochemical experiments where controlled amino acid supplementation is required for studying metabolic activity and cellular processes.
Catalog No. 50-006-61154