Learn More
Medchemexpress LLC L-Aspartic acid potassium | 1115-63-5 | 171.19 | 50 G

Supplier: Medchemexpress LLC HYN0666A50G
L-Aspartic acid potassium is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues, and also in the study of inflammatory conditions.
- Amino acid for various research applications.
- Utilized in preparing prodrugs targeting colon and cecal tissues.
- Applicable in studies related to inflammatory conditions.
- Molecular weight of 171.19.
- Chemical formula: C4H6KNO4.
- Presented as a white to off-white solid.
- Soluble in water at 100 mg/mL (584.15 mM) with sonication.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.