Learn More
Medchemexpress LLC L-aspartic acid sodium | 3792-50-5 | 155.08 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN0666C500MG
Encompass_Preferred
L-Aspartic acid sodium is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues and in the study of inflammatory conditions.
  • Amino acid
  • Used for preparing prodrugs to target colon and cecal tissues
  • Used in the study of inflammatory conditions
Catalog No. 50-003-88860