Learn More
Medchemexpress LLC L-Aspartic acid (Standard) | 56-84-8 | MFCD00002616 | 133.10 | 50 MG
Supplier: Medchemexpress LLC HYN0666R50MG
L-Aspartic acid (Standard) is the analytical standard for L-Aspartic acid, intended for research and analytical applications. It is an amino acid commonly used for preparing prodrugs to target colon and cecal tissues and in studies of inflammatory conditions. It is described as a solid with a white to off-white appearance.
- Analytical standard for L-Aspartic acid
- Used for preparing prodrugs to target colon and cecal tissues
- Used in studies of inflammatory conditions
- Solid with a white to off-white appearance
- Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS