Learn More
Medchemexpress LLC L-Aspartyl-L-phenylalanine | 13433-09-5 | 98.0% | 280.28 | 1 G

Supplier: Medchemexpress LLC HY230931G
L-Aspartyl-L-phenylalanine is a metabolite of aspartame that can inhibit angiotensin converting enzyme (ACE) purified from rabbit lungs with a Ki of 11 μM. This product is for research use only and appears as a white to off-white solid.
- Inhibits angiotensin converting enzyme (ACE)
- Ki of 11 μM against angiotensin converting enzyme
- Molecular weight: 280.28
- Molecular formula: C13H16N2O5
- CAS No.: 13433-09-5
- Stable under recommended storage conditions
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.