Learn More
Cayman Chemical L-Aspartyl-L-phenylalnIn 1g
SDP
Supplier:  Cayman Chemical 341971 G
Encompass_Preferred
An ACE inhibitor and aspartame metabolite; an ACE inhibitor (Ki = 11 µM); serum levels are positively associated with pancreatic ductal adenocarcinoma
Catalog No. 50-294-6420