Learn More
Medchemexpress LLC L-biotin | 21788-37-4 | 99.7% | 244.31 | C10H16N2O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW048449
Encompass_Preferred
L-biotin is the unnatural enantiomer of biotin and is used as a biochemical research reagent. It is provided as a white to off-white solid with reported high purity and has been observed to bind streptavidin in laboratory assays.
  • High purity suitable for research applications.
  • White to off-white solid, easy to handle and store.
  • Soluble in DMSO at high concentration for in vitro use.
  • Available in small solid quantities appropriate for bench-scale experiments.
  • Stable when stored frozen as a powder to extend shelf life.
Catalog No. 50-002-08762