Learn More
Medchemexpress LLC HY-B0399 500mg Medchemexpress, L-Carnitine CAS:541-15-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0399500MG
Encompass_Preferred
Medchemexpress, HY-B0399 500mg L-Carnitine CAS:541-15-1 L-Carnitine (Levocarnitine) is an endogenous molecule involved in fatty acid metabolism, biosynthesized within the human body using amino acids: L-lysine and L-methionine, as substrates. L-Carnitine functions to transport long chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation. L-carnitine can ameliorate metabolic imbalances in many inborn errors of metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2519