Learn More
Medchemexpress LLC L-carnitine-d3 hydrochloride | 350818-62-1 | 98.0% | 200.68 | C7H13D3ClNO3 | 10 MG

Supplier: Medchemexpress LLC HYB1453S10MG
L-Carnitine-d3 hydrochloride is the deuterium-labeled hydrochloride salt of L-carnitine supplied as a research reagent and stable-isotope internal standard for analytical applications. The product is provided in small mg-scale packs, with documented purity, molecular weight, and storage recommendations; safety data sheet and data sheet are available.
- Deuterium-labeled L-carnitine hydrochloride suitable as an internal standard or tracer for research.
- Supplied as a solid, off-white to light yellow.
- Purity 98.0%.
- Molecular weight 200.68 g/mol.
- Available in small pack sizes (5 mg-100 mg).
- Storage: 4°C sealed; in solvent, -80°C (6 months) or -20°C (1 month).
- Safety data sheet and product data sheet available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.