Learn More
Apexbio Technology LLC L-Carnitine inner salt 541-15-1 5g
SDP
Supplier:  Apexbio Technology LLC N19355000
Encompass_Preferred
L-Carnitine inner salt (CAS 541-15-1) is a naturally occurring quaternary ammonium compound prevalent in mammalian tissues particularly in skeletal and cardiac muscle It facilitates the transport of long-chain fatty acids into mitochondria for -oxidation thereby playing a critical role in cellular energy metabolism By modulating substrate utilization L-carnitine influences the balance between glucose and lipid metabolism It is widely employed in metabolic research to study energy regulation mitochondrial function and disorders involving altered fatty acid oxidation
Catalog No. 50-000-90095