Learn More
Medchemexpress LLC L-Carnitine (Standard) | 541-15-1 | MFCD00038747 | 100.0% | 100 MG
Supplier: Medchemexpress LLC HYB0399R100MG
L-Carnitine (Standard) is the analytical standard of L-Carnitine. It is a highly polar, small zwitterion and an essential co-factor for the mitochondrial β-oxidation pathway. This product is intended for research and analytical applications.
- Functions to transport long-chain fatty acyl-CoAs into the mitochondria for degradation by β-oxidation.
- Acts as an antioxidant.
- Can ameliorate metabolic imbalances in many inborn errors of metabolism.
- Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.