Learn More
Medchemexpress LLC L-Carnosine-d4 | 2714339-90-7 | 99.0% | 230.26 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW013494S1MG
Encompass_Preferred
L-Carnosine-d4 is the deuterium-labeled version of L-Carnosine. L-Carnosine is a dipeptide composed of the amino acids beta-alanine and histidine, and it has the potential to mitigate many biochemical changes associated with aging.
  • Deuterium-labeled L-carnosine.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes incorporated into drug molecules can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54741