Learn More
TARGETMOL CHEMICALS INC L-CHICORIC ACID 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6S23915MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. L-Chicoric Acid (trans-Caffeoyltartaric acid) has been shown to inhibit hyaluronidase and HIV-1 integrase and to possess phagoeytosis stimulatory activity in vitro and in vivo and antiviral acitivy. L-Chicoric acid may reduce acute alcohol-induced steatosis in mice through interfering with the induction of iNOS and iNOS-dependent signaling cascades in the liver. 3. L-Chicoric acid inhibited cell viability and induced apoptosis in 3T3-L1 preadipocytes which was characterized by chromatin condensation and poly ADP-ribose-polymerase (PARP) cleavage. purity: 99%
Catalog No. 50-257-6371