Learn More
Medchemexpress LLC L-citronellol | 7540-51-4 | MFCD00063214 | 99.9% | 156.27 | C10H20O | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW015515
Encompass_Preferred
L-citronellol is the (S)-enantiomer of 3,7-dimethyloct-6-en-1-ol supplied as a high-purity analytical standard for laboratory research and analytical chemistry (CAS 7540-51-4). It is used in stereochemical studies, chromatography, and formulation research where the levorotatory form is required.
  • High purity (≈99.9%) for analytical applications.
  • Well-defined stereochemistry as the L-(S)-enantiomer.
  • Suitable as an analytical standard for chromatography and method development.
  • Compatible with storage recommendations for long-term stability.
  • Available in multiple pack sizes to support different scale experiments.
Catalog No. 50-003-04139