Learn More
Medchemexpress LLC L-Citrulline-13C | 94740-46-2 | 176.18 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0391S35MG
Encompass_Preferred
L-Citrulline-13C is the 13C-labeled L-Citrulline, an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from l-arginine via the arginine-citrulline pathway. This compound is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89970