Learn More
Medchemexpress LLC L-Citrulline | 372-75-8 | 175.19 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN039110MM/1ML
Encompass_Preferred
L-Citrulline is an amino acid derived from ornithine in the catabolism of proline or glutamine and glutamate, or from L-arginine via the arginine-citrulline pathway. It is also obtained during the degradation of asymmetric dimethylarginine (ADMA), a process catalyzed by dimethylarginine dimethylaminohydrolase (DDAH), yielding dimethylamine (DMA) as a coproduct. It is an endogenous metabolite and human endogenous metabolite.
  • Derived from ornithine or L-arginine pathways
  • Obtained during ADMA degradation
  • Molecular weight of 175.19
  • For research use only
Catalog No. 50-003-89268