Learn More
Medchemexpress LLC L-citrulline-d7 | 2483831-24-7 | 99.8% | 182.23 | C6H6D7N3O3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN0391S510MG
Encompass_Preferred
L-Citrulline-d7 is the deuterium-labeled isotopologue of L-citrulline supplied as a solid stable-isotope standard for research and analytical applications. It is intended for use as an internal standard or tracer in quantitative mass spectrometry and metabolic studies, and is provided with documented purity and storage recommendations.
  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • Molecular formula C6H6D7N3O3 and molecular weight 182.23.
  • High chemical purity (99.8%).
  • Solid powder form with storage recommendations: powder -20°C (3 years) or 4°C (2 years).
  • Supplied as a 10 mg aliquot suitable for analytical use.
Catalog No. 50-002-95871