Learn More
Medchemexpress LLC L-Cyclohexylalanine | 27527-05-5 | 171.24 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW002450
Encompass_Preferred
L-Cyclohexylalanine is an amino acid derivative that modifies an atrial natriuretic peptide. It regulates body fluid homeostasis, blood pressure, and exhibits vasodilation activity. This product is for research use only and has not been fully validated for medical applications.
  • Amino acid derivative
  • Modifies atrial natriuretic peptide
  • Regulates body fluid and blood pressure homeostasis
  • Exhibits vasodilation activity
Catalog No. 50-004-49405