Learn More
Medchemexpress LLC L-Cyclohexylalanine | 27527-05-5 | 171.24 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00245050G
Encompass_Preferred
L-Cyclohexylalanine is an amino acid derivative that modifies an atrial natriuretic peptide, regulating homeostasis of body fluid and blood pressure, and exhibiting vasodilation activity. This product is for research use only.
  • Amino acid derivative
  • Modifies atrial natriuretic peptide
  • Regulates homeostasis of body fluid and blood pressure
  • Exhibits vasodilation activity
  • Suitable for research applications
  • Related to cardiovascular disease research
Catalog No. 50-004-49576