Learn More
Cayman Chemical L-CystathIonIn 50mg
SDP
Supplier:  Cayman Chemical 1606150 MG
Encompass_Preferred
A precursor of cysteine formed in the methionine transsulfuration pathway; deficiency can indicate the presence of metabolic disorders such as cystathioninuria
Catalog No. 50-297-3068