Learn More
Cayman Chemical L-CystathIonIn 5mg
SDP
Supplier:  Cayman Chemical 160615 MG
Encompass_Preferred
A precursor of cysteine formed in the methionine transsulfuration pathway; deficiency can indicate the presence of metabolic disorders such as cystathioninuria
Catalog No. 50-297-0390