Learn More
Medchemexpress LLC L-Cystathionine dihydrochloride | 61125-50-6 | 295.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW009749C
Encompass_Preferred
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid associated with the metabolic state of sulfur-containing amino acids. It offers protection against Homocysteine-induced mitochondria-dependent apoptosis in vascular endothelial cells (HUVECs) and plays an important role in cardiovascular protection.
  • Nonprotein thioether
  • Key amino acid associated with the metabolic state of sulfur-containing amino acids
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis in vascular endothelial cells (HUVECs)
  • Plays a significant role in cardiovascular protection
  • Stable under recommended storage conditions
  • Store at -20°C, under nitrogen, away from moisture
Catalog No. 50-004-53243