Learn More
Medchemexpress LLC L-Cystathionine (dihydrochloride) | 61125-50-6 | 99.9% | 295.18 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW009749CR1MG
Encompass_Preferred
L-Cystathionine dihydrochloride is a nonprotein thioether and a key amino acid involved in the metabolic state of sulfur-containing amino acids. This white to off-white solid substance is recommended for storage at -20°C in sealed conditions, away from light and moisture, and under nitrogen.
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells
  • Significant in cardiovascular protection
  • Provided as a white to off-white solid
  • Store at -20°C in sealed conditions, away from light and moisture, and under nitrogen
Catalog No. 50-004-53345