Learn More
Medchemexpress LLC L-Cystathionine dihydrochloride | 61125-50-6 | 99.9% | 295.18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW009749C50MG
Encompass_Preferred
L-Cystathionine dihydrochloride (Standard) is the analytical standard of L-Cystathionine dihydrochloride. It is intended for research and analytical applications. This product is a nonprotein thioether and a key amino acid associated with the metabolic state of sulfur-containing amino acids. It protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs) and plays an important role in cardiovascular protection.
  • Analytical standard
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
  • Nonprotein thioether
  • Key amino acid associated with the metabolic state of sulfur-containing amino acids
  • Protects against Homocysteine-induced mitochondria-dependent apoptosis of vascular endothelial cells (HUVECs)
  • Plays an important role in cardiovascular protection
Catalog No. 50-004-53385