Learn More
Medchemexpress LLC L-cysteic acid | 498-40-8 | MFCD00007524 | 98.0% | 169.16 | C3H7NO5S | 5G
SDP
Supplier:  Medchemexpress LLC HY1240095G
Encompass_Preferred
L-cysteic acid is the oxidized derivative of L-cystine used as a research reagent and endogenous metabolite reference. It is reported as a substrate for GADCase/CADCaseII and is used in biochemical studies of sulfur-containing amino acid metabolism and related enzymatic assays. Supplied as a solid with documentation for research use.
Catalog No. 50-002-36273