Learn More
Medchemexpress LLC L-cysteic acid | 498-40-8 | MFCD00007524 | 98.0% | 169.16 g/mol | C3H7NO5S | 10G
SDP
Supplier:  Medchemexpress LLC HY12400910G
Encompass_Preferred
L-cysteic acid (CAS 498-40-8) is the oxidation product of l-cystine and a sulfonic acid-containing amino acid used in biochemical research. It serves as a substrate for GADCase/CADCase II, shows high affinity for CADCase I (Km = 0.22 mM), and is transported by a Na+-dependent synaptosomal uptake system. The compound is supplied as a white to off-white solid with research-grade specifications.
  • High purity 98.0%.
  • Molecular formula C3H7NO5S; molecular weight 169.16 g/mol.
  • Suitable for metabolic and transport studies.
  • Available in solid and pre-made solution formats.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-35889