Learn More
Medchemexpress LLC L-cysteic acid | 498-40-8 | MFCD00007524 | 98.0% | 169.16 g/mol | C3H7NO5S | 1G
SDP
Supplier:  Medchemexpress LLC HY1240091G
Encompass_Preferred
L-cysteic acid is the oxidation product of L-cystine and is used as a biochemical research reagent and endogenous metabolite probe. It functions as a substrate for GADCase/CADCaseII and is taken up by synaptosomes via a high-affinity, Na+-dependent transport system. The product is supplied in both solid and solution formats and includes documented solubility and storage data to support experimental planning.
Catalog No. 50-002-38406