Learn More
Medchemexpress LLC L-cysteic acid | 498-40-8 | MFCD00007524 | 98.0% | 169.16 g/mol | C3H7NO5S | 500MG

Supplier: Medchemexpress LLC HY124009500MG
L-cysteic acid (CAS 498-40-8) is the oxidation product of L-cystine and an endogenous metabolite used in biochemical and neurochemical research. It functions as a substrate for GADCase/CADCase II and is actively taken up by synaptosomes via a high-affinity, Na+-dependent transport system. The compound is provided as a research-grade solid or in solution formats for laboratory assays.
- Oxidation product of L-cystine used in metabolic and enzymatic studies.
- Substrate for GADCase/CADCase II, suitable for enzyme assays.
- Taken up by synaptosomes via a Na+-dependent transport mechanism.
- Molecular formula C3H7NO5S; molecular weight 169.16 g/mol.
- Available as solid powder and solution preparations for lab use.
- Purity typically 98.0% for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.