Learn More
Medchemexpress LLC L-cysteine-1-13C | 224054-24-4 | MFCD09842706 | 99.0% | 122.15 g·mol⁻¹ | C2[13C]H7NO2S | 100mg
SDP
Supplier:  Medchemexpress LLC HYY0337S3100MG
Encompass_Preferred
L-Cysteine-1-13C is the 13C-labeled L-Cysteine L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]
Catalog No. 50-002-69816