Learn More
Medchemexpress LLC L-Cysteine-1,2,3-13C3-15N, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)- | 360795-94-4 | 589.68 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW007798S10MG
Encompass_Preferred
Fmoc-Cys(Trt)-OH-1,2,3-13C3,15N is a 15N-labeled and 13C-labeled Fmoc-Cys(Trt)-OH-1,2,3. It is a cysteine derivative for research use only, not for medical applications. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-47068