Learn More
Medchemexpress LLC L-Cysteine-3-13C | 201612-57-9 | 98.5% | 122.15 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYY0337S2
Encompass_Preferred
L-Cysteine-3-13C is the 13C-labeled L-Cysteine. L-Cysteine is a conditionally essential amino acid, acting as a precursor for biologically active molecules such as hydrogen sulfide (H2S), glutathione, and taurine. It also suppresses ghrelin and reduces appetite in rodents and humans.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-93430