Learn More
Medchemexpress LLC L-cysteine | 52-90-4 | MFCD00064306 | >97.0% | 121.16 g/mol | C3H7NO2S | 1mL
SDP
Supplier:  Medchemexpress LLC HYY033710MM/1ML
Encompass_Preferred
L-Cysteine (Cysteine) is an orally active conditionally essential amino acid with hypoglycemic effects which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine promotes the proliferation and differentiation of neural stem cells via the CBS/H2S pathway L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans L-Cysteine can be used as an anorectic agent[1][2][3][4]
Catalog No. 50-002-69333