Learn More
Medchemexpress LLC L-cysteine | 52-90-4 | MFCD00064306 | 99.9% | 121.16 g/mol | C3H7NO2S | 10mg
SDP
Supplier:  Medchemexpress LLC HYY0337R10MG
Encompass_Preferred
L-Cysteine (Standard) is the analytical standard of L-Cysteine This product is intended for research and analytical applications L-Cysteine is a conditionally essential amino acid which acts as a precursor for biologically active molecules such as hydrogen sulphide (H2S) glutathione and taurine L-Cysteine suppresses ghrelin and reduces appetite in rodents and humans[1]
Catalog No. 50-002-65892