Learn More
Selleck Chemical LLC L-Cysteine HCl S4815-25mg
SDP
Supplier:  Selleck Chemical LLC S481525MG
Encompass_Preferred
L-cysteine Hydrochloride also known as L-CYSTEINE or Cysteine HCL (anhydrous) is classified as a cysteine or a cysteine derivative that increases glutathione levels and is important for lung and brain function and liver detoxification
Catalog No. 50-311-2021