Learn More
Research Products International Corp L-Cysteine Hydrochloride Monohydrate, 100 Grams
SDP
Supplier:  Research Products International Corp C81020100.0
Encompass_Preferred
L-Cysteine Hydrochloride Monohydrate is a compound that is a derivative of the amino acid cysteine and is commonly used as a precursor in the synthesis of proteins and peptides. 
It is often utilized in cell culture media to support the growth and viability of cells. L-Cysteine is an essential component for the synthesis of glutathione, an important antioxidant in cells. 
It is employed in biotechnological processes for the production of recombinant proteins. It can act as a reducing agent, facilitating the proper folding and stability of proteins.  
Researchers may use this chemical in various laboratory experiments and studies, given its role in protein synthesis and cellular processes.
Catalog No. 50-213-264