Learn More
Bioworld L-Cysteine Hydrochloride, 100 g
Supplier:  Bioworld 403001402
Encompass_Preferred
Used in fields of medicine, food processing, biological study, materials of chemical industry and so on. Technical Information:High Purity GradeSpecific rotation: +.5.7 to +6.8°Moisture: 8.0-12.0%Chloride: 19.8-20.8%Sulfate: <0.03%Iron: <0.003%Heavy metals (as Pb): <0.0015%Appearance: White Crystalline PowderDensity : 800 kg/m3HS Code : 29309013IUPAC_Name: L-Cysteine Hydrochloride HydrateMelting Point: 175°CPH Value: 1.0 - 2 at 25 g/lRTECS Number: HA2285000
Catalog No. 50-255-688