Learn More
Medchemexpress LLC L-cysteine hydrochloride hydrate | 7048-04-6 | MFCD00065606 | 95.7% | 175.63 | C3H10ClNO3S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW016715R
Encompass_Preferred
L-Cysteine hydrochloride hydrate is an analytical reference standard of L-cysteine in its hydrochloride hydrate form (CAS 7048-04-6). It is intended for analytical calibration, method development, and biochemical research, and serves as a precursor to hydrogen sulfide, glutathione, and taurine.
  • High purity (95.7%) for reliable analytical results.
  • White to off-white solid appearance for easy handling and visual inspection.
  • Available in small package sizes (25 mg-1 g) suited to method development and standards preparation.
  • Functions as a biochemical precursor useful in metabolic and signaling studies.
  • Certificate of Analysis provides assay, storage, and handling details.
Catalog No. 50-003-02894