Learn More
Alkali Scientific L Cysteine Hydrochloride Monohydrate, 5 Kilograms
SDP
Supplier:  Alkali Scientific CYS004CP
Encompass_Preferred
L Cysteine Hydrochloride Monohydrate, 5 kilograms, is a high-quality amino acid used extensively in biochemical research. This product serves as a source of sulfur for the synthesis of proteins, enzymes, and other molecules in the body. It is commonly used in laboratory experiments focusing on antioxidant activity, enzyme catalysis, and protein structure analysis. L Cysteine is also an important component in the production of glutathione, a major antioxidant in cells. Available in the monohydrate form, it offers stable, easy-to-handle conditions for laboratories that require bulk quantities of this critical amino acid for scientific studies.
Catalog No. 50-006-60827