Learn More
Medchemexpress LLC L-Cysteine methyl ester hydrochloride | 18598-63-5 | MFCD00038985 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB103810MM/1ML
Encompass_Preferred
L-Cysteine methyl ester hydrochloride is an antitussive, phlegmolytic agent used to relieve breathing difficulties caused by large amounts of phlegm. It is also a copper corrosion inhibitor used in various industrial studies.
  • Stimulates the secretory activity of trachea secretory cells.
  • At high concentrations, it stimulates the synthesis of mucinous glycoprotein in submucosal glands.
  • Has an obvious viscosity-reducing effect on secretory cell mucous.
  • Can overcome the harmful effects of morphine on respiration and gas exchange in Sprague Dawley rats without affecting the sedative or early anti-sensory effects of opioids.
Catalog No. 50-004-12439