Learn More
Medchemexpress LLC L-cysteine, N-[(9H-fluoren-9-ylmethoxy)carbonyl] | 135248-89-4 | MFCD03788178 | 98.4% | 343.40 g/mol | C18H17NO4S | 5 G

Supplier: Medchemexpress LLC HYW0487275G
(((9H-fluoren-9-yl)methoxy)carbonyl)-L-cysteine is an Fmoc-protected L-cysteine derivative used as a protected amino acid building block in peptide synthesis and related biochemical applications.
- chemical formula C18H17NO4S.
- molecular weight 343.40 g/mol.
- appearance white to off-white solid.
- purity 98.39% (HPLC) as provided on the certificate of analysis.
- available in multiple pack sizes (for example, 1 g, 5 g, 10 g, 25 g).
- storage: powder at 4°C under nitrogen; in solvent, store at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.