Learn More
Alkali Scientific L Cystine, 1 Kg
SDP
Supplier:  Alkali Scientific YST002CP
Encompass_Preferred
L-Cystine (1 kg) is a sulfur-containing amino acid essential in cellular metabolism and protein structure. This product is widely used in cell culture media and biochemical assays, particularly in studies related to oxidative stress, protein synthesis, and cellular defense mechanisms. L-Cystine plays a crucial role in maintaining the stability of proteins through the formation of disulfide bonds, which are vital for enzyme and protein structure. It is also used in research involving the synthesis of glutathione, a key antioxidant in cells. Additionally, L-Cystine is essential for studying sulfur metabolism and its impact on cell growth and immune function.
Catalog No. 50-006-61787