Learn More
Alkali Scientific L Cystine, 100 G
SDP
Supplier:  Alkali Scientific YST001CP
Encompass_Preferred
L-Cystine (100g) is a sulfur-containing amino acid used in cell culture and biochemical research. L-Cystine plays an important role in cellular processes such as protein synthesis and the formation of disulfide bonds in proteins, contributing to the structural stability of enzymes and other proteins. This product is widely used in the growth media for cell culture and in research studies investigating the role of sulfur-containing amino acids in cellular metabolism. It is especially valuable in the study of oxidative stress, protein folding, and cellular defense mechanisms. L-Cystine is also used in the production of glutathione, a crucial antioxidant in cells.
Catalog No. 50-006-61762