Learn More
Medchemexpress LLC L-Cystine-15N2 | 952407-61-3 | 242.29 | 100 UG

Supplier: Medchemexpress LLC HYN0394S2100UG
L-Cystine-15N2 is an 15N-labeled version of L-Cystine, an amino acid and intracellular thiol. L-Cystine plays a vital role in regulating cellular processes. This product is intended for research use only.
- Purity: 99.27%.
- Applications include use as a tracer.
- Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
- Recommended storage for powder: -20°C for 3 years or 4°C for 2 years.
- Recommended storage in solvent: -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.