Learn More
Medchemexpress LLC L-cystine-3,3'-13C2 | 2483736-13-4 | 99.0% | 242.29 g·mol⁻1 | C4(13C)2H12N2O4S2 | 10 MG

Supplier: Medchemexpress LLC HYN0394S410MG
L-Cystine-3,3'-13C2 is the 13C-labelled form of L-cystine, supplied as a purified solid for use as a stable isotope standard or tracer in biochemical and analytical applications.
- Isotopically labeled at positions 3 and 3' with 13C.
- Purity 99.0%.
- Molecular weight 242.29 g·mol⁻1; molecular formula C4(13C)2H12N2O4S2.
- Provided as a solid in small-quantity vials suitable for analytical use.
- Intended for use as a stable isotope standard, tracer, or reference in mass spectrometry and metabolic studies.
- CAS number 2483736-13-4.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.