Learn More
Medchemexpress LLC L-Cystine-34S2 | 113512-08-6 | MFCD34676341 | 244.11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0394S31MG
Encompass_Preferred
L-Cystine-34S2 is a stable 34S-labeled derivative of the amino acid L-cysteine, which is a disulfide compound. It is typically stored at -20 °C to maintain stability.
  • Stable 34S-labeled derivative of L-cysteine.
  • Disulfide compound.
  • Stored at -20 °C.
Catalog No. 50-003-89126